5-Bromo-2-chlorotoluene ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Bromo-1-chloro-2-methylbenzene, 1-Bromo-4-chloro-3-methylbenzene

A10602.06 A10602.14 A10602.22
ALFAA10602.06EA 0 SGD
ALFAA10602.06 ALFAA10602.14 ALFAA10602.22
5-Bromo-2-chlorotoluene ≥98%
5-Bromo-2-chlorotoluene

Purity: 98% CAS No.: 54932-72-8 Molecular Formula: C7H6BrCl Formula Weight: 205.48 Flash Point: 96°(205°F) Refractive Index: 1.5745 Boiling Point: 90-93°/10mm Beilstein Reference: 3234896 MDL No.: MFCD00018503

Formula: C₇H₆BrCl
Boiling Pt: 90…93 °C (10 mmHg)
Density: 1,549
Flash Pt: 96 °C (205 °F)
Storage Temperature: Ambient
MDL Number: MFCD00018503
CAS Number: 54932-72-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR