D-(+)-Xylose ≥98%

Supplier: Thermo Fisher Scientific

A10643.0E A10643.22 A10643.36
ALFAA10643.0EEA 0 SGD
ALFAA10643.0E ALFAA10643.22 ALFAA10643.36
D-(+)-Xylose ≥98%
D(+)-Xylose

Purity: 98+% CAS No.: 58-86-6 Molecular Formula: C5H10O5 Formula Weight: 150.13 Flash Point: 100°(212°F) Physical Form: Cryst powder Density: 1.525 Sensitivity: Hygroscopic Appearance: White Melting Point: 147-151° Beilstein Reference: 1562108 Merck Reference: 14,10087 Optical Rotation: +19° (c=10 in water + NH4OH) MDL No.: MFCD00151475

Formula: C₅H₁₀O₅
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 147…151 °C
Density: 1,525
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00151475
CAS Number: 58-86-6
EINECS: 200-400-7
Merck Index: 14,10087
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f045c9c134fcd7d287b95738095c40d429a173e&vid=447bd80bb2f3ca1cb52dc711d0957e9ab6e2f84d&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR