2,5-Diphenyloxazole 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: PPO, 2,5-Diphenyl-1,3-oxazole

A10654.14 A10654.22 A10654.36
ALFAA10654.14EA 0 SGD
ALFAA10654.14 ALFAA10654.22 ALFAA10654.36
2,5-Diphenyloxazole 99%
2,5-Diphenyloxazole

Purity: 99% CAS No.: 92-71-7 Molecular Formula: C15H11NO Formula Weight: 221.26 Melting Point: 70-74° Boiling Point: 360° MDL No.: MFCD00005306

Formula: C₁₅H₁₁NO
MW: 221.26 g/mol
Boiling Pt: 360 °C
Melting Pt: 70…74 °C
Density: 1.060
Storage Temperature: Ambient
MDL Number: MFCD00005306
CAS Number: 92-71-7
EINECS: 202-181-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR