ω-Bromo-3-methoxyacetophenone ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Bromo-3'-methoxyacetophenone, Bromomethyl m-methoxyphenyl ketone, 3-Methoxyphenacyl bromide, 2-Bromo-1-(3-methoxyphenyl)ethanone, 2-Bromo-1-(3-methoxyphenyl)ethan-1-one, α-Bromo-3-methoxyacetophenone

A10716.06 A10716.14
ALFAA10716.06EA 0 SGD
ALFAA10716.06 ALFAA10716.14
ω-Bromo-3-methoxyacetophenone ≥98%
ω-Bromo-3-methoxyacetophenone

Purity: 98% CAS No.: 5000-65-7 Molecular Formula: C9H9BrO2 Formula Weight: 229.08 Storage Temperature: 2°C to 8°C Odour: Irritating Beilstein Reference: 510128 UN No.: 3261 MDL No.: MFCD00000199

Formula: C₉H₉BrO₂
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 60…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000199
CAS Number: 5000-65-7
EINECS: 225-666-1
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR