4-Bromo-3-nitrotoluene ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-Bromo-4-methyl-2-nitrobenzene

A10718.14 A10718.22
ALFAA10718.14EA 0 SGD
ALFAA10718.14 ALFAA10718.22
4-Bromo-3-nitrotoluene ≥98%
4-Bromo-3-nitrotoluene

Purity: 98+% CAS No.: 5326-34-1 Molecular Formula: C7H6BrNO2 Formula Weight: 216.04 Flash Point: 110°(230°F) Melting Point: 29-32° Boiling Point: 96°/1mm Beilstein Reference: 1945925 MDL No.: MFCD00024180

Formula: C₇H₆BrNO₂
Boiling Pt: 96 °C (1 mmHg)
Melting Pt: 29…32 °C
Density: 1,578
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00024180
CAS Number: 5326-34-1
EINECS: 226-203-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR