Methyl hexanoate 99

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester

A10757.0B A10757.18 A10757.30
ALFAA10757.0BEA 0 SGD
ALFAA10757.0B ALFAA10757.18 ALFAA10757.30
Methyl hexanoate 99
Methyl hexanoate

Purity: 99% CAS No.: 106-70-7 Molecular Formula: CH3(CH2)4CO2CH3 Formula Weight: 130.19 Flash Point: 45°(113°F) Refractive Index: 1.4050 Melting Point: -71° Boiling Point: 150-151° Beilstein Reference: 1744683 UN No.: 3272 MDL No.: MFCD00009510

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 150…151 °C
Melting Pt: –71 °C
Density: 0.885
Flash Pt: 45 °C (113 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009510
CAS Number: 106-70-7
EINECS: 203-425-1
UN: 3272
ADR: 3 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR