Antimony(III) potassium oxitartrate hydrate ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (R,R)-(+)-Antimony potassium tartrate hydrate, (+)-Antimony potassium L-tartrate hydrate, L(+)-Tartaric acid antimony potassium salt hydrate, L(+)-Antimony potassium tartrate hydrate, Antimony potassium L(+)-tartrate hydrate, (R,R)-(+)-Tartaric acid antimony potassium salt hydrate, Antimony potassium tartrate hydrate, Potassium antimony(III) tartrate hydrate

A10889.22 A10889.0E A10889.36
ALFAA10889.22EA 0 SGD
ALFAA10889.22 ALFAA10889.0E ALFAA10889.36
Antimony(III) potassium oxitartrate hydrate ≥98%
Antimony (III) potassium oxitartrate hydrate

Purity: 98% CAS No.: 331753-56-1 Molecular Formula: C8H4K2O12Sb2.xH2O Formula Weight: 613.82(anhy) Physical Form: Powder Density: 2.607 Appearance: White UN No.: 1551 MDL No.: MFCD00148863

Formula: C₈H₄K₂O₁₂Sb₂·H₂O
MW: 631.84 g/mol (anhy)
Density: 2.607
Storage Temperature: Ambient
MDL Number: MFCD00148863
CAS Number: 331753-56-1
EINECS: 234-293-3
UN: 1551
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR