4-Bromo-2-fluoroanisole ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-bromo-2-fluoro-1-methoxybenzene

A10948.06 A10948.14 A10948.22
ALFAA10948.06EA 0 SGD
ALFAA10948.06 ALFAA10948.14 ALFAA10948.22
4-Bromo-2-fluoroanisole ≥97%
4-Bromo-2-fluoroanisole

Purity: 97% CAS No.: 2357-52-0 Molecular Formula: C7H6BrFO Formula Weight: 205.03 Flash Point: 98°(208°F) Refractive Index: 1.5450 Boiling Point: 208° Beilstein Reference: 3240714 MDL No.: MFCD00011710

Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 208 °C
Melting Pt: 16
Density: 1.590
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011710
CAS Number: 2357-52-0
EINECS: 219-096-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR