Furfuryl alcohol ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Furfurol

A10968.18 A10968.30 A10968.0B
ALFAA10968.18EA 51.5 SGD
ALFAA10968.18 ALFAA10968.30 ALFAA10968.0B
Furfuryl alcohol ≥98%
Furfuryl alcohol

Purity: 98% CAS No.: 98-00-0 Molecular Formula: C5H6O2 Formula Weight: 98.10 Flash Point: 65°(149°F) Refractive Index: 1.4870 Odour: Mild Melting Point: -31° Boiling Point: 170-171° Beilstein Reference: 106291 Merck Reference: 14,4305 UN No.: 2874 MDL No.: MFCD00003252

Formula: C₅H₆O₂
Boiling Pt: 170…171 °C
Melting Pt: –31 °C
Density: 1,129
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003252
CAS Number: 98-00-0
EINECS: 202-626-1
UN: 2874
ADR: 6.1,III
Merck Index: 14,4305

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR