2-Bromo-5-fluorotoluene ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Bromo-4-fluoro-2-methylbenzene

A11112.14 A11112.22 A11112.06
ALFAA11112.14EA 0 SGD
ALFAA11112.14 ALFAA11112.22 ALFAA11112.06
2-Bromo-5-fluorotoluene ≥98%
2-Bromo-5-fluorotoluene

Purity: 98+% CAS No.: 452-63-1 Molecular Formula: C7H6BrF Formula Weight: 189.03 Flash Point: 70°(158°F) Refractive Index: 1.5260 Boiling Point: 169-170° Beilstein Reference: 1859121 MDL No.: MFCD00017921

Formula: C₇H₆BrF
Boiling Pt: 169…170 °C
Density: 1,495
Flash Pt: 70 °C (158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017921
CAS Number: 452-63-1
EINECS: 207-203-5
UN: 1993
ADR: 3,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR