Ethyl formate ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Formic acid ethyl ester

A11113.0F A11113.AE A11113.AP
ALFAA11113.0FEA 0 SGD
ALFAA11113.0F ALFAA11113.AE ALFAA11113.AP
Ethyl formate ≥97%
Ethyl formate

Purity: 97% CAS No.: 109-94-4 Molecular Formula: C3H6O2 Formula Weight: 74.08 Storage Temperature: 2°C to 8°C Flash Point: -19°(-2°F) Physical Form: Liquid Density: 0.921 Refractive Index: 1.3600 Appearance: Colourless Odour: Fruit-like pH: 4.1 Melting Point: -81° Boiling Point: 52-54° Beilstein Reference: 906769 Merck Reference: 14,3807 UN No.: 1190 MDL No.: MFCD00003294

Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 52…54 °C
Melting Pt: –81 °C
Density: 0.921
Flash Pt: –19 °C (–2 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003294
CAS Number: 109-94-4
EINECS: 203-721-0
UN: 1190
ADR: 3 II
Merck Index: 14,3807

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR