(±)-1,1'-Bi-2-naphthol ≥99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1,1'-Binaphthyl-2,2'-Diol, 2,2'-Dihydroxy-1,1'-binaphthyl, 1,1'-Bi-2-naphthol, [1,1'-Binaphthalene]-2,2'-diol

A11182.06 A11182.14 A11182.22
ALFAA11182.06EA 0 SGD
ALFAA11182.06 ALFAA11182.14 ALFAA11182.22
(±)-1,1'-Bi-2-naphthol ≥99%
(±)-1,1'-Bi-2-naphthol

Purity: 99% CAS No.: 602-09-5 Molecular Formula: C20H14O2 Formula Weight: 286.33 Odour: Characteristic Melting Point: 215-218° Beilstein Reference: 997518 Merck Reference: 14,1226 UN No.: 2811 MDL No.: MFCD00004068

Formula: C₂₀H₁₄O₂
Melting Pt: 215…218 °C
Density: 1,301
Storage Temperature: Ambient
MDL Number: MFCD00004068
CAS Number: 602-09-5
EINECS: 210-014-0
UN: 2811
ADR: 6.1,III
Merck Index: 14,1226

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR