Ethyl laurate 98

Supplier: Thermo Fisher Scientific

Synonyms: Dodecanoic acid ethyl ester, Ethyl dodecanoate, Ethyl Laurate, Dodecylic acid ethyl ester, Lauric acid ethyl ester

A11188.0F A11188.AE A11188.AP
ALFAA11188.0FEA 0 SGD
ALFAA11188.0F ALFAA11188.AE ALFAA11188.AP
Ethyl laurate 98
Ethyl laurate

Purity: 98% CAS No.: 106-33-2 Molecular Formula: CH3(CH2)10CO2CH2CH3 Formula Weight: 228.38 Flash Point: 138°(280°F) Refractive Index: 1.4315 Odour: Fruit-like Melting Point: -2° Boiling Point: 268-270° Beilstein Reference: 1769671 Merck Reference: 14,3818 MDL No.: MFCD00015065

Formula: C₁₄H₂₈O₂
MW: 228.38 g/mol
Boiling Pt: 268…270 °C
Melting Pt: –2 °C
Density: 0.862
Flash Pt: 138 °C (280 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015065
CAS Number: 106-33-2
EINECS: 203-386-0
Merck Index: 13,03851

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR