Ethyl difluoroacetate ≥98%

Supplier: Thermo Fisher Scientific
Danger

A11215.06 A11215.14 A11215.22
ALFAA11215.06EA 0 SGD
ALFAA11215.06 ALFAA11215.14 ALFAA11215.22
Ethyl difluoroacetate ≥98%
Ethyl difluoroacetate

Purity: 98% CAS No.: 454-31-9 Molecular Formula: F2CHCO2CH2CH3 Formula Weight: 124.09 Flash Point: 25°(77°F) Refractive Index: 1.3475 Boiling Point: 98-99° Beilstein Reference: 1748597 UN No.: 2924 MDL No.: MFCD00013578

Formula: C₄H₆F₂O₂
Boiling Pt: 98…99 °C
Density: 1,242
Flash Pt: 25 °C (77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013578
CAS Number: 454-31-9
EINECS: 207-223-4
ADR: 3,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR