2-Bromo-4-fluorophenol ≥98%

Supplier: Thermo Fisher Scientific
Warning

A11263.06 A11263.14 A11263.22
ALFAA11263.06EA 0 SGD
ALFAA11263.06 ALFAA11263.14 ALFAA11263.22
2-Bromo-4-fluorophenol ≥98%
2-Bromo-4-fluorophenol

Purity: 98+% CAS No.: 496-69-5 Molecular Formula: C6H4BrFO Formula Weight: 191.00 Flash Point: 85°(185°F) Melting Point: 42-46° Boiling Point: 123-125°/50mm Beilstein Reference: 2355593 MDL No.: MFCD00010614

Formula: C₆H₄BrFO
Boiling Pt: 123…125 °C (50 mmHg)
Melting Pt: 42…46 °C
Flash Pt: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010614
CAS Number: 496-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR