Ethyl sorbate ≥97%

Supplier: Thermo Fisher Scientific
Warning

A11431.14 A11431.22 A11431.36
ALFAA11431.14EA 0 SGD
ALFAA11431.14 ALFAA11431.22 ALFAA11431.36
Ethyl sorbate ≥97%
Ethyl sorbate

Purity: 98% CAS No.: 2396-84-1 Molecular Formula: CH3CH=CHCH=CHCO2CH2CH3 Formula Weight: 140.18 Flash Point: 69°(156°F) Refractive Index: 1.4950 Boiling Point: 195-196° Beilstein Reference: 1362941 MDL No.: MFCD00009296

Formula: C₈H₁₂O₂
Boiling Pt: 195…196 °C
Density: 0,948
Flash Pt: 69 °C (156 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009296
CAS Number: 2396-84-1
EINECS: 219-258-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR