p-Toluamide 98

Supplier: Thermo Fisher Scientific

Synonyms: 4-Methylbenzamide

A11559.14 A11559.22 A11559.36
ALFAA11559.14EA 0 SGD
ALFAA11559.14 ALFAA11559.22 ALFAA11559.36
p-Toluamide 98
p-Toluamide

Purity: 98+% CAS No.: 619-55-6 Molecular Formula: C8H9NO Formula Weight: 135.17 Melting Point: 158-160° Beilstein Reference: 774353

Formula: C₈H₉NO
MW: 135.17
Boiling Pt: 305
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00008002
CAS Number: 619-55-6
EINECS: 210-600-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR