2-Fluoro-6-iodobenzonitrile ≥98%

Supplier: Thermo Fisher Scientific
Danger

A11576.06
ALFAA11576.06EA 0 SGD
ALFAA11576.06
2-Fluoro-6-iodobenzonitrile ≥98%
2-Fluoro-6-iodobenzonitrile

Purity: 98% CAS No.: 79544-29-9 Molecular Formula: C7H3FIN Formula Weight: 247.01 Melting Point: 48-52° Beilstein Reference: 4971873 UN No.: 3439 MDL No.: MFCD00015478

Formula: C₇H₃FIN
Boiling Pt: 281 °C (1013 hPa)
Melting Pt: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00015478
CAS Number: 79544-29-9
UN: 3439
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR