1-Bromo-4-fluorobenzene ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Bromofluorobenzene

A11630.0E A11630.22 A11630.36
ALFAA11630.0EEA 911 SGD
ALFAA11630.0E ALFAA11630.22 ALFAA11630.36
1-Bromo-4-fluorobenzene ≥99%
1-Bromo-4-fluorobenzene

Purity: 99% CAS No.: 460-00-4 Molecular Formula: C6H4BrF Formula Weight: 175.01 Flash Point: 60°(140°F) Refractive Index: 1.5270 Odour: Aromatic Melting Point: -16° Boiling Point: 151-153° Beilstein Reference: 774102 UN No.: 1993 MDL No.: MFCD00000342

Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 151…153 °C
Melting Pt: –16 °C
Density: 1.604
Flash Pt: 60 °C (140 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000342
CAS Number: 460-00-4
EINECS: 207-300-2
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR