Bis(trimethylsilyl)carbodiimide ≥97%

Supplier: Thermo Fisher Scientific
Warning

A11670.06 A11670.14
ALFAA11670.06EA 0 SGD
ALFAA11670.06 ALFAA11670.14
Bis(trimethylsilyl)carbodiimide ≥97%
Bis(trimethylsilyl)carbodiimide

Purity: 97% CAS No.: 1000-70-0 Molecular Formula: C7H18N2Si2 Formula Weight: 186.41 Flash Point: 42°(107°F) Refractive Index: 1.4340 Odour: Mild Melting Point: -23° Boiling Point: 163-164° Beilstein Reference: 2042082 UN No.: 1993 MDL No.: MFCD00051538

Formula: C₇H₁₈N₂Si₂
Boiling Pt: 163…164 °C
Melting Pt: –23 °C
Density: 0,823
Flash Pt: 42 °C (107 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051538
CAS Number: 1000-70-0
EINECS: 213-673-2
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR