Diboron trioxide ≥99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Boron oxide, Boric anhydride

A11707.36 A11707.0E
ALFAA11707.36EA 0 SGD
ALFAA11707.36 ALFAA11707.0E
Diboron trioxide ≥99%
di-Boron trioxide

Purity: 99% CAS No.: 1303-86-2 Molecular Formula: B2O3 Formula Weight: 69.62 Odour: Odourless Melting Point: ca 450° Boiling Point: 1860° Merck Reference: 14,1337 MDL No.: MFCD00011315

Formula: B₂O₃
MW: 69.62 g/mol
Boiling Pt: 1860 °C
Melting Pt: ∼450 °C
Density: 2.46
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS Number: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 13,01327

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR