Trityl chloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Trt-Cl, Chlorotriphenylmethane, Triphenylmethyl chloride, (Chloromethanetriyl)tribenzene, Triphenylchloromethane

A11799.14 A11799.22 A11799.36
ALFAA11799.14EA 0 SGD
ALFAA11799.14 ALFAA11799.22 ALFAA11799.36
Trityl chloride ≥98%
Trityl chloride

Purity: 98% CAS No.: 76-83-5 Molecular Formula: C19H15Cl Formula Weight: 278.78 Melting Point: 110-114° Boiling Point: 230-235°/20mm UN No.: 3261 MDL No.: MFCD00000813

Formula: C₁₉H₁₅Cl
MW: 278.78 g/mol
Boiling Pt: 230…235 °C (20 mmHg)
Melting Pt: 110…114 °C
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000813
CAS Number: 76-83-5
EINECS: 200-986-4
UN: 3261
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR