2-Hydroxy-6-methylnicotinic acid 98

Supplier: Thermo Fisher Scientific

Synonyms: 2-Hydroxy-6-methylpyridine-3-carboxylic acid

A11842.06 A11842.14
ALFAA11842.06EA 0 SGD
ALFAA11842.06 ALFAA11842.14
2-Hydroxy-6-methylnicotinic acid 98
2-Hydroxy-6-methylnicotinic acid

Purity: 98+% CAS No.: 38116-61-9 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: 233-238° Beilstein Reference: 129164

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 233…238 °C
Storage Temperature: Ambient
MDL Number: MFCD00006272
CAS Number: 38116-61-9
EINECS: 253-784-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR