3-Bromo-o-toluidine ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Amino-6-bromotoluene, 3-Bromo-2-methylaniline

A11884.06 A11884.14
ALFAA11884.06EA 0 SGD
ALFAA11884.06 ALFAA11884.14
3-Bromo-o-toluidine ≥98%
3-Bromo-o-toluidine

Purity: 98+% CAS No.: 55289-36-6 Molecular Formula: C7H8BrN Formula Weight: 186.06 Flash Point: 110°(230°F) Refractive Index: 1.6160 Boiling Point: 253-255° Beilstein Reference: 2354213 UN No.: 2810 MDL No.: MFCD00051579

Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 253…255 °C
Density: 1.510
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051579
CAS Number: 55289-36-6
EINECS: 259-568-5
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR