Ethyl-5-bromovalerate 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Bromovaleric acid ethyl ester, Ethyl 5-bromovalerate

A11952.22 A11952.14 A11952.36
ALFAA11952.22EA 0 SGD
ALFAA11952.22 ALFAA11952.14 ALFAA11952.36
Ethyl-5-bromovalerate 98
Ethyl 5-bromovalerate

Purity: 98% CAS No.: 14660-52-7 Molecular Formula: C7H13BrO2 Formula Weight: 209.09 Flash Point: 103°(217°F) Refractive Index: 1.4580 Boiling Point: 227° Beilstein Reference: 1752023 MDL No.: MFCD00000266

Formula: BrH₂C(CH₂)₃COOCH₂CH₃
MW: 209.08 g/mol
Boiling Pt: 227 °C
Density: 1.306
Flash Pt: 103 °C (217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000266
CAS Number: 14660-52-7
EINECS: 238-705-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR