N-(Chloromethyl)phthalimide ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(Chloromethyl)isoindoline-1,3-dione

A12018.14 A12018.22
ALFAA12018.14EA 0 SGD
ALFAA12018.14 ALFAA12018.22
N-(Chloromethyl)phthalimide ≥97%
N-(Chloromethyl)phthalimide

Purity: 97% CAS No.: 17564-64-6 Molecular Formula: C9H6ClNO2 Formula Weight: 195.61 Melting Point: 131-135° Beilstein Reference: 140942

Formula: C₉H₆ClNO₂
MW: 195.6 g/mol
Boiling Pt: 305
Melting Pt: 131…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005898
CAS Number: 17564-64-6
EINECS: 241-541-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR