4-Bromo-2-(trifluoromethyl)aniline ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Bromo-α,α,α-trifluoro-o-toluidine, 2-Amino-5-bromobenzotrifluoride, 2-Amino-5-bromo-α,α,α-trifluorotoluene, 2-Amino-5-bromo-α,α,α-trifluortoluene

A12146.06 A12146.14
ALFAA12146.06EA 0 SGD
ALFAA12146.06 ALFAA12146.14
4-Bromo-2-(trifluoromethyl)aniline ≥98%
4-Bromo-2-(trifluoromethyl)aniline

Purity: 98% CAS No.: 445-02-3 Molecular Formula: C7H5BrF3N Formula Weight: 240.03 Flash Point: 107°(224°F) Refractive Index: 1.5320 Boiling Point: 84-85°/5mm Beilstein Reference: 2211504 UN No.: 2810 MDL No.: MFCD00064393

Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 84…85 °C (5 mmHg)
Density: 1.710
Flash Pt: 107 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064393
CAS Number: 445-02-3
EINECS: 207-150-8
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR