1-Bromo-4-chlorobenzene ≥98%

Supplier: Thermo Fisher Scientific
Danger

A12160.0B A12160.22 A12160.30
ALFAA12160.0BEA 0 SGD
ALFAA12160.0B ALFAA12160.22 ALFAA12160.30
1-Bromo-4-chlorobenzene ≥98%
1-Bromo-4-chlorobenzene

Purity: 98+% CAS No.: 106-39-8 Molecular Formula: C6H4BrCl Formula Weight: 191.46 Flash Point: 100°(212°F) Odour: Aromatic Boiling Point: 196° Beilstein Reference: 1099014 UN No.: 2811 MDL No.: MFCD00000600

Formula: C₆H₄BrCl
Boiling Pt: 196 °C
Melting Pt: 63…67 °C
Density: 1,651
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000600
CAS Number: 106-39-8
EINECS: 203-392-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR