1-Bromo-4-heptylbenzene ≥97%

Supplier: Thermo Fisher Scientific

Synonyms: (4-Bromophenyl)heptane

A12235.03 A12235.06 A12235.14
ALFAA12235.03EA 0 SGD
ALFAA12235.03 ALFAA12235.06 ALFAA12235.14
1-Bromo-4-heptylbenzene ≥97%
1-Bromo-4-heptylbenzene

Purity: 97% CAS No.: 76287-49-5 Molecular Formula: C13H19Br Formula Weight: 255.20 MDL No.: MFCD00061115

Formula: C₁₃H₁₉Br
MW: 255.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061115
CAS Number: 76287-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR