Ethyl-4-pentenoate ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Ethyl pent-4-enoate, 4-Pentenoic Acid Ethyl Ester

A12422.03 A12422.06 A12422.14 A12422.22
ALFAA12422.03EA 0 SGD
ALFAA12422.03 ALFAA12422.06 ALFAA12422.14 ALFAA12422.22
Ethyl-4-pentenoate ≥98%
Ethyl-4-pentenoate

Purity: 98+% CAS No.: 1968-40-7 Molecular Formula: CH2=CHCH2CH2CO2CH2CH3 Formula Weight: 128.17 Flash Point: 37°(98°F) Refractive Index: 1.4170 Boiling Point: 146° Beilstein Reference: 1751643 UN No.: UN3272 MDL No.: MFCD00036562

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 146 °C
Density: 0.900
Flash Pt: 37 °C (98 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036562
CAS Number: 1968-40-7
EINECS: 217-818-0
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR