Potassium hydrogen sulphate 97

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Potassium bisulphate

A12428.0C A12428.0E A12428.22 A12428.36
ALFAA12428.0CEA 0 SGD
ALFAA12428.0C ALFAA12428.0E ALFAA12428.22 ALFAA12428.36
Potassium hydrogen sulphate 97
Potassium hydrogen sulphate

Purity: 97% CAS No.: 7646-93-7 Molecular Formula: HKO4S Formula Weight: 136.17 Odour: Odourless Melting Point: 195-197° dec. UN No.: 2509 MDL No.: MFCD00011404

Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 195…197 °C (dec.)
Density: 2.240
Storage Temperature: Ambient
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8 II
Merck Index: 13,07695

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR