2,5-Diiodothiophene ≥99%

Supplier: Thermo Fisher Scientific
Warning

A12443.06 A12443.14
ALFAA12443.06EA 0 SGD
ALFAA12443.06 ALFAA12443.14
2,5-Diiodothiophene ≥99%
2,5-Diiodothiophene

Purity: 99% CAS No.: 625-88-7 Molecular Formula: C4H2I2S Formula Weight: 335.93 Storage Temperature: 2°C to 8°C Melting Point: 39-43° Boiling Point: 139-140°/15mm Beilstein Reference: 109915 MDL No.: MFCD00014525

Formula: C₄H₂I₂S
MW: 335.93 g/mol
Boiling Pt: 139…140 °C (15 mmHg)
Melting Pt: 39…43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014525
CAS Number: 625-88-7
EINECS: 210-915-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR