5-Fluoro-2-nitrophenol ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Fluoro-2-hydroxynitrobenzene

A12568.09 A12568.18 A12568.30
ALFAA12568.09EA 0 SGD
ALFAA12568.09 ALFAA12568.18 ALFAA12568.30
5-Fluoro-2-nitrophenol ≥98%
5-Fluoro-2-nitrophenol

Purity: 98% CAS No.: 446-36-6 Molecular Formula: C6H4FNO3 Formula Weight: 157.10 Flash Point: 91°(195°F) Melting Point: 30-36° Boiling Point: 260° Beilstein Reference: 1870311 MDL No.: MFCD00007107

Formula: C₆H₄FNO₃
MW: 157.1 g/mol
Boiling Pt: 260 °C
Melting Pt: 30…36 °C
Flash Pt: 91 °C (195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007107
CAS Number: 446-36-6
EINECS: 207-168-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR