ω-Bromo-4-chloroacetophenone ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Bromo-4'-chloroacetophenone, 2-Bromo-1-(4-chlorophenyl)ethanone, Bromomethyl p-chlorophenyl ketone, α-Bromo-4-chloroacetophenone, 2-Bromo-4′-chloroacetophenone, 4-Chlorophenacyl bromide

A12607.14 A12607.22 A12607.36
ALFAA12607.14EA 0 SGD
ALFAA12607.14 ALFAA12607.22 ALFAA12607.36
ω-Bromo-4-chloroacetophenone ≥98%
ω-Bromo-4-chloroacetophenone

Purity: 98% CAS No.: 536-38-9 Molecular Formula: C8H6BrClO Formula Weight: 233.50 Melting Point: 94-98° Beilstein Reference: 607603 Merck Reference: 14,2153 UN No.: 3261 MDL No.: MFCD00000625

Formula: C₈H₆BrClO
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00000625
CAS Number: 536-38-9
EINECS: 208-631-5
UN: 3261
ADR: 8,III
Merck Index: 14,2153

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR