1-Bromo-4-pentyloxybenzene ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: 1-Bromo-4-(pentyloxy)benzene, 1-Bromo-4-pentoxy-benzene

A12614.06
ALFAA12614.06EA 0 SGD
ALFAA12614.06
1-Bromo-4-pentyloxybenzene ≥98%
1-Bromo-4-pentyloxybenzene

CAS No.: 30752-18-2 Molecular Formula: C11H15BrO Formula Weight: 243.15 MDL No.: MFCD00156123

Formula: C₁₁H₁₅BrO
MW: 243.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00156123
CAS Number: 30752-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR