Dichlorophen 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2,2'-Methylenebis(4-chlorophenol), Bis(5-chloro-2-hydroxyphenyl)methane

A12640.22 A12640.14 A12640.36
ALFAA12640.22EA 0 SGD
ALFAA12640.22 ALFAA12640.14 ALFAA12640.36
Dichlorophen 95%
Dichlorophen

Purity: 95% CAS No.: 97-23-4 Molecular Formula: C13H10Cl2O2 Formula Weight: 269.13 Melting Point: 172-176° Beilstein Reference: 1884514 Merck Reference: 14,3071 UN No.: 3077 MDL No.: MFCD00002322

Formula: C₁₃H₁₀Cl₂O₂
Boiling Pt: 389 °C (1013 hPa)
Melting Pt: 172…176 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002322
CAS Number: 97-23-4
EINECS: 202-567-1
ADR: 9,III
Merck Index: 14,3071

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR