Ethyl butyrylacetate ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethyl Butyrylacetate, Ethyl 3-oxohexanoate

A12786.0B A12786.18 A12786.30
ALFAA12786.0BEA 0 SGD
ALFAA12786.0B ALFAA12786.18 ALFAA12786.30
Ethyl butyrylacetate ≥98%
Ethyl butyrylacetate

Purity: 98% CAS No.: 3249-68-1 Molecular Formula: C8H14O3 Formula Weight: 158.20 Flash Point: 78°(172°F) Refractive Index: 1.4270 Melting Point: -44° Boiling Point: 94-95°/12mm Beilstein Reference: 507689 MDL No.: MFCD00009401

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
Boiling Pt: 94…95 °C (12 mmHg)
Melting Pt: –44 °C
Density: 0.990
Flash Pt: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009401
CAS Number: 3249-68-1
EINECS: 221-835-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR