6-Amino-3-bromo-2-methylpyridine 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 6-Amino-3-bromo-2-picoline, 6-AMINO-3-BROMO-2-METHYLPYRIDINE, 3-Bromo-2-methyl-6-pyridylamine, 5-Bromo-6-methylpyridin-2-amine, 3-Bromo-2-methylpyridin-6-amine, 2-Amino-5-bromo-6-picoline, ['6-Amino-3-bromo-2-picoline'], 5-Bromo-6-methyl-2-pyridylamine, 2-Amino-5-bromo-6-methylpyridine

A12948.03 A12948.06 A12948.14
ALFAA12948.03EA 0 SGD
ALFAA12948.03 ALFAA12948.06 ALFAA12948.14
6-Amino-3-bromo-2-methylpyridine 97%
6-Amino-3-bromo-2-methylpyridine

Purity: 97% CAS No.: 42753-71-9 Molecular Formula: C6H7BrN2 Formula Weight: 187.04 Melting Point: 80-82° Beilstein Reference: 114140

Formula: C₆H₇BrN₂
Melting Pt: 80…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00068230
CAS Number: 42753-71-9
EINECS: 255-927-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR