4-Bromo-2-nitrotoluene 99

Supplier: Thermo Fisher Scientific
Danger

A12951.06 A12951.22
ALFAA12951.06EA 0 SGD
ALFAA12951.06 ALFAA12951.22
4-Bromo-2-nitrotoluene 99
4-Bromo-2-nitrotoluene

Purity: 99% CAS No.: 60956-26-5 Molecular Formula: C7H6BrNO2 Formula Weight: 216.04 Flash Point: 110°(230°F) Melting Point: 45-48° Boiling Point: 130°/12mm Beilstein Reference: 2328190 MDL No.: MFCD00041243

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 130 °C (12 mmHg)
Melting Pt: 45…48 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041243
CAS Number: 60956-26-5
EINECS: 262-536-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR