1-Bromo-4-ethylbenzene ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-(Bromophenyl)ethane

A13013.09 A13013.18 A13013.30
ALFAA13013.09EA 0 SGD
ALFAA13013.09 ALFAA13013.18 ALFAA13013.30
1-Bromo-4-ethylbenzene ≥99%
1-Bromo-4-ethylbenzene

Purity: 99% CAS No.: 1585-07-5 Molecular Formula: C8H9Br Formula Weight: 185.06 Flash Point: 63°(145°F) Refractive Index: 1.5440 Boiling Point: 203-204° Beilstein Reference: 1856989 MDL No.: MFCD00000111

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 203…204 °C
Melting Pt: –43.5 °C
Density: 1.341
Flash Pt: 63 °C (145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000111
CAS Number: 1585-07-5
EINECS: 216-439-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR