Cinnamyl alcohol 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Cinnamic alcohol, 3-Phenylprop-2-en-1-ol

A13025.0B A13025.30
ALFAA13025.0BEA 0 SGD
ALFAA13025.0B ALFAA13025.30
Cinnamyl alcohol 98
Cinnamyl alcohol

Purity: 98% CAS No.: 104-54-1 Molecular Formula: C9H10O Formula Weight: 134.18 Flash Point: 126°(258°F) Odour: Pleasant Boiling Point: 257-258° Beilstein Reference: 1903999 Merck Reference: 14,2302 MDL No.: MFCD00002921

Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 257…258 °C
Melting Pt: 32…35 °C
Density: 1.044
Flash Pt: 126 °C (258 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002921
CAS Number: 104-54-1
EINECS: 203-212-3
Merck Index: 13,02325

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR