α-Bromo-2-nitrotoluene ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: α-Bromo-o-nitrotoluene, 1-(Bromomethyl)-2-nitrobenzene, alpha-Bromo-2-nitrotoluene, 2-Nitrobenzyl bromide

A13127.06 A13127.14 A13127.22
ALFAA13127.06EA 0 SGD
ALFAA13127.06 ALFAA13127.14 ALFAA13127.22
α-Bromo-2-nitrotoluene ≥98%
α-Bromo-2-nitrotoluene

Purity: 98+% CAS No.: 3958-60-9 Molecular Formula: C7H6BrNO2 Formula Weight: 216.04 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 45-48° Beilstein Reference: 638991 UN No.: 3261 MDL No.: MFCD00007184

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 45…48 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007184
CAS Number: 3958-60-9
EINECS: 223-558-9
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR