4-Hydroxybenzyl alcohol 99

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (4-Hydroxyphenyl)methanol, p-Hydroxybenzyl Alcohol, 4-(Hydroxymethyl)phenol

A13132.18 A13132.22 A13132.30
ALFAA13132.18EA 0 SGD
ALFAA13132.18 ALFAA13132.22 ALFAA13132.30
4-Hydroxybenzyl alcohol 99
4-Hydroxybenzyl alcohol

Purity: 99% CAS No.: 623-05-2 Molecular Formula: C7H8O2 Formula Weight: 124.14 Melting Point: 114-122° Boiling Point: 251-253° MDL No.: MFCD00004658

Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 251…253 °C
Melting Pt: 114…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00004658
CAS Number: 623-05-2
EINECS: 210-768-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR