Ethyl 4-hydroxybenzoate 99

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate

A13172.30 A13172.0B A13172.0I
ALFAA13172.30EA 0 SGD
ALFAA13172.30 ALFAA13172.0B ALFAA13172.0I
Ethyl 4-hydroxybenzoate 99
Ethyl 4-hydroxybenzoate

Purity: 99% CAS No.: 120-47-8 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 116-118° Boiling Point: 297-298° dec. Merck Reference: 14,3837 MDL No.: MFCD00002353

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 297…298 °C (dec.)
Melting Pt: 116…118 °C
Density: 1.078 g/cm³ (131 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002353
CAS Number: 120-47-8
EINECS: 204-399-4
Merck Index: 13,03869

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR