4-Bromobenzyl alcohol 99

Supplier: Thermo Fisher Scientific

Synonyms: (4-Bromophenyl)methanol

A13197.09 A13197.18
ALFAA13197.09EA 0 SGD
ALFAA13197.09 ALFAA13197.18
4-Bromobenzyl alcohol 99
4-Bromobenzyl alcohol

Purity: 99% CAS No.: 873-75-6 Molecular Formula: C7H7BrO Formula Weight: 187.04 Melting Point: 76-79° Beilstein Reference: 1931620

Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 268
Melting Pt: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00004650
CAS Number: 873-75-6
EINECS: 212-851-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR