3-Methyl-2-thiophenecarbonitrile 96%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-Methylthiophene-2-carbonitrile

A13305.06 A13305.03 A13305.14
ALFAA13305.06EA 0 SGD
ALFAA13305.06 ALFAA13305.03 ALFAA13305.14
3-Methyl-2-thiophenecarbonitrile 96%
3-Methyl-2-thiophenecarbonitrile

Purity: 96% CAS No.: 55406-13-8 Molecular Formula: C6H5NS Formula Weight: 123.18 Refractive Index: 1.5530 Boiling Point: 91-93°/12mm UN No.: 3276 MDL No.: MFCD00052086

Formula: C₆H₅NS
MW: 123.18 g/mol
Boiling Pt: 91…93 °C (12 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00052086
CAS Number: 55406-13-8
UN: 3276
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR