2,4,5-Trifluorobenzonitrile ≥98%

Supplier: Thermo Fisher Scientific
Danger

A13332.14
ALFAA13332.14EA 0 SGD
ALFAA13332.14
2,4,5-Trifluorobenzonitrile ≥98%
2,4,5-Trifluorobenzonitrile

Purity: 98+% CAS No.: 98349-22-5 Molecular Formula: C7H2F3N Formula Weight: 157.10 Flash Point: 83°(181°F) Refractive Index: 1.4736 Boiling Point: 168-170° Beilstein Reference: 5427045 UN No.: 3276 MDL No.: MFCD00013289

Formula: C₇H₂F₃N
Boiling Pt: 168…170 °C
Density: 1,373
Flash Pt: 83 °C (181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013289
CAS Number: 98349-22-5
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR