2,4-Dichlorobenzyl alcohol ≥99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: (2,4-Dichlorophenyl)methanol

A13338.14 A13338.22 A13338.36
ALFAA13338.14EA 0 SGD
ALFAA13338.14 ALFAA13338.22 ALFAA13338.36
2,4-Dichlorobenzyl alcohol ≥99%
2,4-Dichlorobenzyl alcohol

Purity: 99% CAS No.: 1777-82-8 Molecular Formula: C7H6Cl2O Formula Weight: 177.03 Flash Point: 110°(230°F) Melting Point: 57-61° Beilstein Reference: 1448652 Merck Reference: 14,3060 MDL No.: MFCD00004606

Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 150 °C (25 mmHg)
Melting Pt: 57…61 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004606
CAS Number: 1777-82-8
EINECS: 217-210-5
Merck Index: 13,03085

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR