2-Bromo-1-indanol ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Bromo-1-hydroxyindane

A13511.18 A13511.09
ALFAA13511.18EA 0 SGD
ALFAA13511.18 ALFAA13511.09
2-Bromo-1-indanol ≥99%
2-Bromo-1-indanol

Purity: 99% CAS No.: 5400-80-6 Molecular Formula: C9H9BrO Formula Weight: 213.08 Melting Point: 128-132° Beilstein Reference: 2046877

Formula: C₉H₉BrO
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00003798
CAS Number: 5400-80-6
EINECS: 226-442-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR