Ethyl piperazine-1-carboxylate ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Ethoxycarbonyl piperazine, Ethyl N-piperazinecarboxylate

A13573.22
ALFAA13573.22EA 0 SGD
ALFAA13573.22
Ethyl piperazine-1-carboxylate ≥99%
Ethyl piperazine-1-carboxylate

Purity: 99% CAS No.: 120-43-4 Molecular Formula: C7H14N2O2 Formula Weight: 158.20 Flash Point: 128°(262°F) Refractive Index: 1.4780 Boiling Point: 237° Beilstein Reference: 125780 MDL No.: MFCD00005964

Formula: C₇H₁₄N₂O₂
MW: 158.2 g/mol
Boiling Pt: 237 °C
Density: 1.080
Flash Pt: 128 °C (262 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005964
CAS Number: 120-43-4
EINECS: 204-395-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR